Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000617.2 | NP_000608.1 | 10, 11 | 1140 | ||
NM_001174125.1 | NP_001167596.1 | 10, 11 | 1132 | |||
NM_001174126.1 | NP_001167597.1 | 10, 11 | 1140 | |||
NM_001174127.1 | NP_001167598.1 | 10, 11 | 1135 | |||
NM_001174128.1 | NP_001167599.1 | 10, 11 | 1135 | |||
NM_001174129.1 | NP_001167600.1 | 10, 11 | 1400 | |||
NM_001174130.1 | NP_001167601.1 | 9, 10 | 1115 | |||
NR_033421.1 | 10, 11 | 1060 | ||||
NR_033422.1 | 10, 11 | 1135 | ||||
XM_005268911.3 | XP_005268968.1 | 10, 11 | 1192 | |||
XM_005268912.4 | XP_005268969.1 | 9, 10 | 1317 | |||
XM_011538404.2 | XP_011536706.1 | 10, 11 | 1516 | |||
XM_011538405.2 | XP_011536707.1 | 10, 11 | 1124 | |||
XM_017019355.1 | XP_016874844.1 | 10, 11 | 1400 | |||
XM_017019356.1 | XP_016874845.1 | 10, 11 | 1330 | |||
GenBank mRNA | AB004857.1 | - | 10, 11 | 1083 | ||
AB062284.1 | - | 10, 11 | 1057 | |||
AF046997.1 | - | 10, 11 | 1093 | |||
AF064484.1 | - | 10, 11 | 1083 | |||
AJ493662.1 | - | 10, 11 | 1132 | |||
AK094735.1 | - | 10, 11 | 1400 | |||
AK128640.1 | - | 10, 11 | 1032 | |||
AK296445.1 | - | 9, 10 | 1094 | |||
AK315987.1 | - | 10, 11 | 1046 | |||
AK316507.1 | - | 9, 10 | 1071 | |||
BC002592.2 | - | 10, 11 | 1087 | 3160522 | ||
BC100014.1 | - | 10, 11 | 1108 | 5300266 | ||
CK000292.1 | - | 6, 7 | 591 | |||
KT455015.1 | - | 10, 11 | 1096 | |||
L37347.1 | - | 7, 8 | 833 |