Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006277.2 | NP_006268.2 | 18 | 2230 | ||
NM_019595.3 | NP_062541.3 | 17 | 2149 | |||
NM_147152.2 | NP_671494.2 | 18 | 2230 | |||
GenBank mRNA | AB033082.1 | - | 17 | 2056 | ||
AF182198.1 | - | 18 | 2209 | |||
AF182199.1 | - | 18 | 2209 | |||
AF248540.1 | - | 16 | 1930 | |||
AK021545.1 | - | 10 | 1137 | |||
BC146779.1 | - | 17 | 2056 | 9007231 |