Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247996.1 | NP_001234925.1 | 12 | 1193 | ||
NM_018482.3 | NP_060952.2 | 11 | 1091 | |||
XM_005250925.1 | XP_005250982.1 | 11 | 1091 | |||
XM_006716563.3 | XP_006716626.1 | 11 | 910 | |||
XM_006716564.1 | XP_006716627.1 | 12 | 1069 | |||
XM_006716565.3 | XP_006716628.1 | 10 | 906 | |||
XM_006716566.1 | XP_006716629.1 | 11 | 1091 | |||
XM_011517052.2 | XP_011515354.1 | 11 | 907 | |||
XM_011517053.1 | XP_011515355.1 | 11 | 1113 | |||
XM_017013467.1 | XP_016868956.1 | 9 | 928 | |||
XM_017013468.1 | XP_016868957.1 | 10 | 1152 | |||
XM_017013469.1 | XP_016868958.1 | 11 | 1091 | |||
GenBank mRNA | AB033075.1 | - | 4 | 315 | ||
BC137135.1 | - | 10 | 888 | 9021132 | ||
BC144493.1 | - | 11 | 990 | |||
BX537768.1 | - | 4 | 344 |