Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316919.1 | NP_001303848.1 | 5, 6 | 1686 | ||
NM_001316920.1 | NP_001303849.1 | 5, 6 | 467 | |||
NM_001316921.1 | NP_001303850.1 | 5, 6 | 756 | |||
NM_001316922.1 | NP_001303851.1 | 5, 6 | 681 | |||
NM_001316923.1 | NP_001303852.1 | 4, 5 | 616 | |||
NM_016078.5 | NP_057162.4 | 5, 6 | 761 | |||
GenBank mRNA | AF151906.1 | - | 5, 6 | 761 | ||
AF223467.1 | - | 5, 6 | 761 | |||
AK128119.1 | - | 2, 3 | 2121 | |||
AK290813.1 | - | 5, 6 | 485 | |||
BC008430.1 | - | 5, 6 | 487 | 4292664 | ||
BQ433266.1 | - | 4, 5 | 382 | |||
BX537929.1 | - | 5, 6 | 477 | |||
BX648167.1 | - | 5, 6 | 1686 | |||
CN390782.1 | - | 4, 5 | 459 | |||
DA067248.1 | - | 5, 6 | 467 | |||
DA773045.1 | - | 4, 5 | 341 |