Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077180.2 | NP_001070648.1 | 2 | 543 | ||
NM_001288659.1 | NP_001275588.1 | 2 | 200 | |||
NM_001288660.1 | NP_001275589.1 | 2 | 200 | |||
NM_016025.4 | NP_057109.3 | 2 | 543 | |||
GenBank mRNA | AF151839.1 | - | 2 | 207 | ||
AJ278577.1 | - | 2 | 461 | |||
AJ278578.1 | - | 2 | 461 | |||
AK074529.1 | - | 2 | 442 | |||
AK075022.1 | - | 2 | 429 | |||
AK075237.1 | - | 2 | 412 | |||
AK313077.1 | - | 2 | 427 | |||
BC000195.1 | - | 2 | 428 | 3350598 | ||
BG183771.1 | - | 2 | 199 | |||
BM149762.1 | - | 2 | 485 | |||
CF146931.1 | - | 2 | 549 |