Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127582.1 | NP_001121054.1 | 4 | 394 | ||
NM_001127583.1 | NP_001121055.1 | 4 | 394 | |||
NM_001127584.1 | NP_001121056.1 | 4 | 394 | |||
NM_001127585.1 | NP_001121057.1 | 3 | 322 | |||
NM_001127586.1 | NP_001121058.1 | 4 | 394 | |||
NM_016162.3 | NP_057246.2 | 4 | 394 | |||
XM_005253698.3 | XP_005253755.1 | 5 | 642 | |||
XM_011520964.2 | XP_011519266.1 | 3 | 323 | |||
XM_011520965.2 | XP_011519267.1 | 4 | 341 | |||
XM_017019392.1 | XP_016874881.1 | 5 | 642 | |||
XM_017019393.1 | XP_016874882.1 | 5 | 642 | |||
GenBank mRNA | AB197695.1 | - | 4 | 353 | ||
AB197696.1 | - | 4 | 353 | |||
AB197697.1 | - | 4 | 353 | |||
AF063594.1 | - | 4 | 372 | |||
AF110645.1 | - | 4 | 370 | |||
AF156552.1 | - | 4 | 370 | |||
AK293169.1 | - | 3 | 316 | |||
AK303354.1 | - | 4 | 376 | |||
AK313621.1 | - | 4 | 366 | |||
BC007781.2 | - | 4 | 356 | 4309278 | ||
BC013038.2 | - | 4 | 339 | |||
BC095434.1 | - | 4 | 358 | 30365729 | ||
BG705367.1 | - | 4 | 394 | |||
BG718170.1 | - | 4 | 405 | |||
BM922497.1 | - | 5 | 623 | |||
BP303219.1 | - | 4 | 394 | |||
EF152349.1 | - | 3 | 281 | |||
EF152350.1 | - | 3 | 186 | |||
EF152351.1 | - | 4 | 353 | |||
EF152352.1 | - | 4 | 353 |