Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286682.1 | NP_001273611.1 | 5 | 551 | ||
NM_001286683.1 | NP_001273612.1 | 5 | 624 | |||
NM_016228.3 | NP_057312.1 | 4 | 757 | |||
NM_182662.1 | NP_872603.1 | 5 | 539 | |||
XM_006714231.2 | XP_006714294.1 | 6 | 681 | |||
XM_011532020.2 | XP_011530322.1 | 2 | 314 | |||
GenBank mRNA | AF481738.1 | - | 4 | 758 | ||
AK055952.1 | - | 5 | 539 | |||
AK314729.1 | - | 5 | 626 | |||
BC031068.1 | - | 5 | 544 | 5286180 |