Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083899.2 | NP_001077368.2 | 3 | 250 | ||
NM_001256017.2 | NP_001242946.2 | 3 | 250 | |||
NM_016363.5 | NP_057447.5 | 3 | 250 | |||
GenBank mRNA | AB035073.1 | - | 3 | 234 | ||
AB043819.1 | - | 3 | 250 | |||
AB043820.1 | - | 3 | 250 | |||
AB043821.1 | - | 3 | 250 | |||
BC104832.1 | - | 3 | 242 | 8143835 | ||
BC111963.1 | - | 3 | 242 | 8327431 |