Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK16 SIRNA Assays ›
Gene Symbol:
CDK16
Gene Name:
cyclin dependent kinase 16
Gene Aliases:
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
Chromosome Location:
Chr.X: 47217881 - 47229997 on Build GRCh38
UniGene ID:
Hs.496068
Species:
Human
siRNA ID
HSS181964
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170460.1
NP_001163931.1
7
972
NM_006201.4
NP_006192.1
7
1107
NM_033018.3
NP_148978.2
7
1214
XM_011543920.2
XP_011542222.1
7
984
XM_011543921.1
XP_011542223.1
8
1379
XM_011543922.1
XP_011542224.1
8
1367
XM_011543923.1
XP_011542225.1
7
1297
XM_011543924.1
XP_011542226.1
7
1107
XM_011543925.2
XP_011542227.1
8
1189
XM_011543926.1
XP_011542228.1
7
2150
XM_011543927.2
XP_011542229.1
8
1227
XM_011543928.1
XP_011542230.1
8
1582
XM_017029569.1
XP_016885058.1
8
1379
XM_017029570.1
XP_016885059.1
8
1189
XM_017029571.1
XP_016885060.1
7
2150
XM_017029572.1
XP_016885061.1
8
1227
XM_017029573.1
XP_016885062.1
8
1582
GenBank mRNA
AB209465.1
-
6
913
AK096881.1
-
7
1096
AK131486.1
-
6
1377
AK290145.1
-
7
1031
AK296880.1
-
7
1008
AK301832.1
-
7
972
AK303849.1
-
8
966
BC001048.2
-
7
1110
3504276
BC006190.2
-
7
924
3830397
BC009852.2
-
5
370
BC015607.2
-
7
1077
4762081
BT006827.1
-
6
672
X66363.1
-
7
795
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
exocytosis
spermatogenesis
growth hormone secretion
neuron projection development
regulation of cell cycle
regulation of insulin secretion involved in cellular response to glucose stimulus
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0