Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 2 | 229 | ||
NM_177937.2 | NP_808800.1 | 2 | 221 | |||
GenBank mRNA | AF236056.1 | - | 2 | 170 | ||
AK027717.1 | - | 2 | 192 | |||
AK075542.1 | - | 2 | 163 | |||
AK075557.1 | - | 2 | 133 | |||
AK092943.1 | - | 2 | 169 | |||
AL533767.3 | - | 2 | 228 | |||
AY358593.1 | - | 2 | 134 | |||
BC001740.1 | - | 2 | 144 | 3351316 | ||
BQ216766.1 | - | 188 | ||||
CR457201.1 | - | 1 | 20 | |||
DA116177.1 | - | 2 | 222 | |||
EU794616.1 | - | 2 | 188 |