Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GMPR2 SIRNA Assays ›
Gene Symbol:
GMPR2
Gene Name:
guanosine monophosphate reductase 2
Gene Aliases:
GMPR 2
Chromosome Location:
Chr.14: 24232417 - 24239242 on Build GRCh38
UniGene ID:
Hs.368855
Species:
Human
siRNA ID
HSS181977
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GMPR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002000.2
NP_001002000.1
3
645
NM_001002001.2
NP_001002001.1
3
586
NM_001002002.2
NP_001002002.1
3
325
NM_001283021.1
NP_001269950.1
3
586
NM_001283022.1
NP_001269951.1
2
566
NM_001283023.1
NP_001269952.1
3
325
NM_016576.4
NP_057660.2
2
566
NR_104265.1
3
645
XM_005267740.3
XP_005267797.1
3
451
XM_005267741.3
XP_005267798.1
3
510
XM_005267742.3
XP_005267799.1
3
329
XM_006720165.2
XP_006720228.1
3
294
XM_017021356.1
XP_016876845.1
3
510
XM_017021357.1
XP_016876846.1
3
549
XM_017021358.1
XP_016876847.1
3
650
XM_017021359.1
XP_016876848.1
3
293
GenBank mRNA
AB032903.1
-
2
192
AF086919.1
-
3
355
AF135159.1
-
3
531
AF419346.1
-
3
435
AK226166.1
-
3
342
AV660418.1
-
3
304
BC003053.1
-
2
515
3546620
BC008021.1
-
2
514
3537989
BC009832.2
-
3
243
3958714
BC093039.1
-
3
384
30347740
BI259066.1
-
2
528
BP347823.1
-
3
325
BX161436.1
-
3
566
BX161518.1
-
3
432
BX247993.1
-
3
248
BX375671.2
-
2
537
CR457156.1
-
2
184
Back To Top
More Information
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleobase metabolic process
purine-containing compound salvage
GMP metabolic process
oxidation-reduction process
GMP reductase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0