Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016589.3 | NP_057673.2 | 2 | 425 | ||
GenBank mRNA | AF139077.1 | - | 2 | 398 | ||
AK026250.1 | - | 2 | 394 | |||
AK225954.1 | - | 2 | 405 | |||
AL136622.1 | - | 2 | 400 | |||
AL390090.1 | - | 2 | 352 | |||
AL390094.1 | - | 2 | 59 | |||
AY358633.1 | - | 2 | 378 | |||
BC012341.1 | - | 2 | 319 | 4576562 | ||
BG705703.1 | - | 2 | 474 | |||
CR533524.1 | - | 2 | 212 |