Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AMZ2 SIRNA Assays ›
Gene Symbol:
AMZ2
Gene Name:
archaelysin family metallopeptidase 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 68205486 - 68257164 on Build GRCh38
UniGene ID:
Hs.293560
Species:
Human
siRNA ID
HSS181997
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AMZ2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033569.1
NP_001028741.1
3
309
NM_001033570.1
NP_001028742.1
3
305
NM_001033571.1
NP_001028743.1
3
281
NM_001033572.1
NP_001028744.1
3
277
NM_001033574.1
NP_001028746.1
2
862
NM_001289054.1
NP_001275983.1
3
289
NM_001289056.1
NP_001275985.1
3
380
NM_016627.4
NP_057711.3
2
862
XM_005257432.4
XP_005257489.1
2
913
XM_005257433.3
XP_005257490.1
3
311
XM_005257435.4
XP_005257492.1
3
253
XM_005257436.3
XP_005257493.1
3
640
XM_006721940.2
XP_006722003.1
3
241
XM_017024723.1
XP_016880212.1
3
500
XM_017024724.1
XP_016880213.1
3
249
XM_017024725.1
XP_016880214.1
2
815
XM_017024726.1
XP_016880215.1
3
276
XM_017024727.1
XP_016880216.1
3
277
XM_017024728.1
XP_016880217.1
2
344
GenBank mRNA
AJ635358.1
-
1
160
AK090981.1
-
1
1148
AK126146.1
-
2
1292
BC050709.1
-
3
241
6060235
BC056271.1
-
3
274
4102397
BF978559.1
-
309
BM450564.1
-
3
250
BQ432895.1
-
1
149
CN278872.1
-
3
275
CN278874.1
-
3
277
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
metallopeptidase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0