Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002602.3 | NP_002593.1 | 2 | 251 | ||
XM_006721942.3 | XP_006722005.1 | 2 | 1224 | |||
XM_017024734.1 | XP_016880223.1 | 2 | 526 | |||
XM_017024735.1 | XP_016880224.1 | 2 | 257 | |||
XM_017024736.1 | XP_016880225.1 | 2 | 191 | |||
GenBank mRNA | BC106884.2 | - | 2 | 208 | 40029486 | |
BQ638010.1 | - | 2 | 192 | |||
BQ639132.1 | - | 2 | 251 | |||
BX537414.1 | - | 2 | 258 | |||
M36476.1 | - | 2 | 208 |