Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 4 | 382 | ||
NM_001283036.1 | NP_001269965.1 | 3 | 292 | |||
NM_001283037.1 | NP_001269966.1 | 3 | 292 | |||
NM_016407.4 | NP_057491.2 | 3 | 292 | |||
XM_017027872.1 | XP_016883361.1 | 6 | 485 | |||
GenBank mRNA | AF117231.1 | - | 3 | 240 | ||
AF161513.1 | - | 3 | 249 | |||
AF161518.1 | - | 3 | 218 | |||
AF212244.1 | - | 3 | 229 | |||
AK000481.1 | - | 3 | 222 | |||
AK000586.1 | - | 2 | 112 | |||
AK026118.1 | - | 3 | 220 | |||
AK225135.1 | - | 3 | 210 | |||
AK293662.1 | - | 3 | 220 | |||
AK302029.1 | - | 4 | 325 | |||
AK314563.1 | - | 3 | 247 | |||
BC003359.1 | - | 3 | 219 | 2900332 | ||
BG468509.1 | - | 3 | 253 | |||
BP307634.1 | - | 3 | 286 | |||
BU537939.1 | - | 3 | 202 | |||
CR983692.1 | - | 3 | 203 | |||
DC397043.1 | - | 4 | 382 |