Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077394.1 | NP_001070862.1 | 4 | 438 | ||
NM_001077395.1 | NP_001070863.1 | 4 | 420 | |||
NM_015958.2 | NP_057042.2 | 4 | 420 | |||
XM_005270938.1 | XP_005270995.1 | 4 | 420 | |||
XM_005270939.4 | XP_005270996.1 | 3 | 228 | |||
XM_011541569.1 | XP_011539871.1 | 4 | 420 | |||
XM_011541570.2 | XP_011539872.1 | 3 | 220 | |||
XM_017001460.1 | XP_016856949.1 | 4 | 440 | |||
XM_017001461.1 | XP_016856950.1 | 3 | 227 | |||
GenBank mRNA | AF132964.1 | - | 4 | 389 | ||
AF248965.1 | - | 4 | 389 | |||
AK125485.1 | - | 5 | 789 | |||
AK225469.1 | - | 4 | 398 | |||
AK225707.1 | - | 4 | 398 | |||
AK225951.1 | - | 4 | 398 | |||
AK289351.1 | - | 4 | 409 | |||
BC053857.1 | - | 4 | 386 | 5503038 | ||
BI907367.1 | - | 4 | 393 | |||
CK002046.1 | - | 4 | 441 | |||
DA133565.1 | - | 4 | 420 |