Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000926.4 | NP_000917.3 | 8 | 3409 | ||
NM_001202474.3 | NP_001189403.1 | 8 | 2659 | |||
NM_001271161.2 | NP_001258090.1 | 7 | 2353 | |||
NM_001271162.1 | NP_001258091.1 | 8 | 1329 | |||
NR_073141.2 | 8 | 2607 | ||||
NR_073142.2 | 7 | 2490 | ||||
NR_073143.2 | 6 | 2222 | ||||
GenBank mRNA | AB084248.1 | - | 7 | 2360 | ||
AB085683.1 | - | 6 | 2229 | |||
AB085843.1 | - | 7 | 2497 | |||
AB085844.1 | - | 7 | 2256 | |||
AB085845.1 | - | 8 | 2614 | |||
AF016381.1 | - | 8 | 2841 | |||
AK304853.1 | - | 8 | 1329 | |||
M15716.1 | - | 8 | 2841 |