Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000294.2 | NP_000285.1 | 6 | 722 | ||
NM_001172432.1 | NP_001165903.1 | 6 | 722 | |||
GenBank mRNA | AB209055.1 | - | 1 | 111 | ||
AK289492.1 | - | 6 | 586 | |||
AK293551.1 | - | 6 | 722 | |||
BC002541.2 | - | 6 | 582 | 3138927 | ||
M14503.1 | - | 1 | 128 | |||
M31606.1 | - | 6 | 605 |