Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004569.3 | NP_004560.1 | 2 | 366 | ||
XM_011536838.2 | XP_011535140.1 | 2 | 364 | |||
XM_017021371.1 | XP_016876860.1 | 2 | 368 | |||
XM_017021372.1 | XP_016876861.1 | 2 | 359 | |||
GenBank mRNA | AK293584.1 | - | 2 | 352 | ||
AK314108.1 | - | 2 | 333 | |||
BC004100.2 | - | 2 | 317 | 3936489 | ||
BC071849.1 | - | 2 | 314 | 5935599 | ||
BP244435.1 | - | 2 | 366 | |||
BT006804.1 | - | 2 | 269 | |||
L19783.1 | - | 2 | 329 |