Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170747.1 | NP_001164218.1 | 3 | 294 | ||
NM_006223.3 | NP_006214.2 | 3 | 294 | |||
NR_033187.1 | 2 | 220 | ||||
GenBank mRNA | AB009690.1 | - | 3 | 215 | ||
AF143096.1 | - | 3 | 208 | |||
AK127605.1 | - | 3 | 205 | |||
AV709926.2 | - | 3 | 243 | |||
BC005234.1 | - | 3 | 213 | |||
BC070288.1 | - | 3 | 213 | |||
BC093700.1 | - | 3 | 291 | 7939545 | ||
BC104653.1 | - | 3 | 214 | |||
BC111394.1 | - | 3 | 217 | |||
BC112281.1 | - | 3 | 291 | 8327749 |
Gene Symbol: PIN4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003571.1 | 1 | 2111 | |||
GenBank mRNA | AK055299.1 | - | 1 | 2111 |