Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKM SIRNA Assays ›
Gene Symbol:
PKM
Gene Name:
pyruvate kinase, muscle
Gene Aliases:
CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1
Chromosome Location:
Chr.15: 72199029 - 72231624 on Build GRCh38
UniGene ID:
Hs.534770
Species:
Human
siRNA ID
HSS182328
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PKM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001206796.2
NP_001193725.1
9
1946
NM_001206797.2
NP_001193726.1
7
1387
NM_001206798.2
NP_001193727.1
8
1634
NM_001206799.1
NP_001193728.1
8
1276
NM_001316318.1
NP_001303247.1
8
1829
NM_002654.5
NP_002645.3
8
1609
NM_182470.3
NP_872270.1
8
1825
NM_182471.3
NP_872271.1
8
1609
XM_005254443.1
XP_005254500.1
9
1333
XM_005254445.4
XP_005254502.1
8
1170
XM_006720570.1
XP_006720633.1
9
1430
XM_011521670.2
XP_011519972.1
8
1395
XM_017022313.1
XP_016877802.1
8
1170
GenBank mRNA
AK092369.1
-
8
1359
AK222927.1
-
8
1175
AK294315.1
-
9
1435
AK297951.1
-
953
AK299413.1
-
8
1113
AK300800.1
-
8
1200
AK307855.1
-
9
1177
AK308264.1
-
8
1200
AK309781.1
-
8
1278
AK312253.1
-
8
1175
BC000481.2
-
8
1191
2964687
BC007640.1
-
8
1175
3533876
BC007952.2
-
8
1187
4299213
BC012811.2
-
8
1187
2958817
BC019265.2
-
3
523
BC023328.1
-
4
559
BC023592.2
-
8
1137
BC035198.1
-
8
1141
3859987
BC094767.1
-
5
736
5296639
BC096823.1
-
8
1376
EU794631.1
-
8
1328
M23725.1
-
8
1196
M26252.1
-
8
1176
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
liver development
ATP biosynthetic process
response to nutrient
response to gravity
programmed cell death
response to muscle inactivity
organ regeneration
response to insulin
pyruvate biosynthetic process
skeletal muscle tissue regeneration
canonical glycolysis
cell-cell adhesion
magnesium ion binding
pyruvate kinase activity
protein binding
ATP binding
kinase activity
MHC class II protein complex binding
potassium ion binding
ADP binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0