Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
siRNA ID
HSS182379
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
5
763
NM_001324418.1
NP_001311347.1
5
766
NM_001324419.1
NP_001311348.1
5
763
NM_001324420.1
NP_001311349.1
5
766
NM_001324421.1
NP_001311350.1
5
766
NM_004194.4
NP_004185.1
5
766
NM_016351.5
NP_057435.2
5
766
NM_021721.4
NP_068367.1
5
766
NM_021722.5
NP_068368.2
5
766
NM_021723.4
NP_068369.1
5
766
XM_005250445.3
XP_005250502.1
4
614
XM_006716028.3
XP_006716091.1
4
614
XM_006716029.3
XP_006716092.1
4
614
XM_011516318.1
XP_011514620.1
5
658
XM_011516319.2
XP_011514621.1
5
742
XM_011516320.2
XP_011514622.1
4
614
XM_011516321.2
XP_011514623.1
4
614
XM_011516322.1
XP_011514624.1
5
658
XM_011516323.1
XP_011514625.1
5
658
XM_011516324.1
XP_011514626.1
5
330
XM_017012329.1
XP_016867818.1
6
709
XM_017012330.1
XP_016867819.1
6
709
XM_017012331.1
XP_016867820.1
5
739
XM_017012332.1
XP_016867821.1
5
658
XM_017012333.1
XP_016867822.1
5
658
XM_017012334.1
XP_016867823.1
6
709
XM_017012335.1
XP_016867824.1
6
709
XM_017012336.1
XP_016867825.1
5
658
XM_017012337.1
XP_016867826.1
6
709
XM_017012338.1
XP_016867827.1
5
737
XM_017012339.1
XP_016867828.1
6
709
XM_017012341.1
XP_016867830.1
4
355
GenBank mRNA
AB009671.1
-
5
564
AF073291.2
-
5
489
AF155381.1
-
5
489
AF155382.1
-
5
489
AF158637.1
-
4
326
AK125432.1
-
4
629
AK291010.1
-
5
613
BC036029.1
-
5
658
BC062433.1
-
5
597
BC092465.1
-
5
658
BC125118.1
-
5
520
40118904
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0