Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 15 | 2653 | ||
NM_001322003.1 | NP_001308932.1 | 15 | 2648 | |||
NM_001322004.1 | NP_001308933.1 | 14 | 2513 | |||
NM_001322005.1 | NP_001308934.1 | 15 | 2765 | |||
NM_001322006.1 | NP_001308935.1 | 14 | 2497 | |||
NM_001322007.1 | NP_001308936.1 | 14 | 2550 | |||
NM_001322008.1 | NP_001308937.1 | 13 | 2410 | |||
NM_001322009.1 | NP_001308938.1 | 15 | 2793 | |||
NM_001322010.1 | NP_001308939.1 | 13 | 2357 | |||
NM_001322011.1 | NP_001308940.1 | 15 | 2604 | |||
NM_001322012.1 | NP_001308941.1 | 15 | 2599 | |||
NM_001322013.1 | NP_001308942.1 | 14 | 2480 | |||
NM_001322014.1 | NP_001308943.1 | 15 | 2686 | |||
NM_001322015.1 | NP_001308944.1 | 15 | 2823 | |||
NR_136154.1 | 15 | 2610 | ||||
XM_006715744.3 | XP_006715807.1 | 8 | 1729 | |||
XM_017012342.1 | XP_016867831.1 | 14 | 2478 | |||
GenBank mRNA | AB103083.1 | - | 11 | 1363 | ||
AB103086.1 | - | 12 | 2248 | |||
AI433648.1 | - | 1 | 177 | |||
AK294661.1 | - | 14 | 2503 | |||
AK312390.1 | - | 15 | 2602 | |||
BC031832.1 | - | 15 | 2617 | |||
BC093921.1 | - | 15 | 2588 | 7939766 | ||
BQ050630.1 | - | 3 | 347 | |||
DB527312.1 | - | 2 | 305 | |||
U14658.1 | - | 15 | 2568 |
Gene Symbol: PMS2CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002217.1 | 6 | 1506 | |||
GenBank mRNA | AB116525.1 | - | 6 | 1506 | ||
BC041364.1 | - | 13 | 2261 |