Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006239.2 | NP_006230.2 | 12, 13 | 1864 | ||
XM_011532039.2 | XP_011530341.1 | 11, 12 | 2097 | |||
GenBank mRNA | AF023456.1 | - | 12, 13 | 1874 | ||
AF023457.1 | - | 11, 12 | 1507 | |||
BX648754.1 | - | 13, 14 | 2440 |