Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002703.4 | NP_002694.3 | 3 | 538 | ||
XM_011534387.1 | XP_011532689.1 | 3 | 315 | |||
GenBank mRNA | AK222759.1 | - | 3 | 477 | ||
AK290942.1 | - | 3 | 393 | |||
AK312297.1 | - | 3 | 514 | |||
BC004200.2 | - | 3 | 410 | 3533162 | ||
D13757.1 | - | 3 | 500 | |||
DB039655.1 | - | 3 | 538 | |||
U00238.1 | - | 3 | 429 |