Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005038.2 | NP_005029.1 | 4 | 462 | ||
GenBank mRNA | AK313929.1 | - | 4 | 463 | ||
BC014893.1 | - | 4 | 449 | |||
BC030707.2 | - | 4 | 458 | 4819748 | ||
BG699755.1 | - | 4 | 467 | |||
CR542081.1 | - | 4 | 350 | |||
L11667.1 | - | 4 | 449 |