Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
siRNA ID
HSS182676
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
12
2216
NM_001162952.2
NP_001156424.1
4
746
NM_001162953.2
NP_001156425.1
12
2219
NM_001289608.1
NP_001276537.1
13
2025
NM_001289609.1
NP_001276538.1
4
746
NM_001289610.1
NP_001276539.1
5
430
NM_032943.4
NP_116561.1
13
2395
NM_206929.3
NP_996812.1
6
550
NM_206930.3
NP_996813.1
3
723
XM_005274057.1
XP_005274114.1
12
5855
XM_005274060.1
XP_005274117.1
13
2039
XM_005274061.1
XP_005274118.1
13
2036
XM_005274062.2
XP_005274119.1
14
2460
XM_005274063.1
XP_005274120.1
12
1919
XM_005274066.1
XP_005274123.1
11
1868
XM_005274067.4
XP_005274124.1
5
2162
XM_011545105.1
XP_011543407.1
13
5903
XM_011545106.1
XP_011543408.1
13
5900
XM_011545107.2
XP_011543409.1
14
6324
XM_011545108.1
XP_011543410.1
12
5783
XM_011545109.2
XP_011543411.1
13
5885
XM_011545110.1
XP_011543412.1
13
5903
XM_011545114.2
XP_011543416.1
4
2068
XM_011545115.1
XP_011543417.1
2
162
XM_011545116.2
XP_011543418.1
3
1947
XM_017017933.1
XP_016873422.1
12
5852
XM_017017934.1
XP_016873423.1
14
6321
XM_017017935.1
XP_016873424.1
10
5699
XM_017017936.1
XP_016873425.1
10
5397
XM_017017937.1
XP_016873426.1
13
1940
XM_017017938.1
XP_016873427.1
12
1898
XM_017017939.1
XP_016873428.1
6
2206
XM_017017940.1
XP_016873429.1
5
2158
GenBank mRNA
AB046817.1
-
13
2406
AJ303364.1
-
1
42
AK024872.1
-
3
887
AK074737.1
-
6
507
AK092121.1
-
3
541
AK124754.1
-
6
2131
AK127504.1
-
7
3094
AK131365.1
-
6
1023
AK222707.1
-
3
887
AK222710.1
-
3
887
AK296224.1
-
13
2007
AK296740.1
-
10
1955
AK298604.1
-
5
348
AL601023.1
-
5
403
AY386362.1
-
6
4410
BC015540.2
-
3
217
BC068495.1
-
4
471
BC110315.1
-
4
275
BC136450.1
-
12
2151
9020439
BC144114.1
-
12
2151
9052634
BC144115.1
-
12
2148
9052635
BX647671.1
-
7
2846
BX648068.1
-
6
2292
CB046277.1
-
6
507
EU035829.1
-
12
1872
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0