Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017846.4 | NP_060316.1 | 4, 5 | 380 | ||
NR_003109.1 | 5, 6 | 528 | ||||
XM_017001542.1 | XP_016857031.1 | 3, 4 | 554 | |||
GenBank mRNA | AF118082.2 | - | 1, 2 | 791 | ||
AK000510.1 | - | 4, 5 | 312 | |||
AK023555.1 | - | 4, 5 | 960 | |||
AK300162.1 | - | 5, 6 | 704 | |||
AK309481.1 | - | 5, 6 | 460 | |||
BC000680.2 | - | 4, 5 | 304 | 3349210 | ||
BC035855.1 | - | 3, 4 | 904 | |||
BC039879.1 | - | 5, 6 | 446 | 5769935 | ||
BI602122.1 | - | 4, 5 | 385 |