Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 11 | 1339 | ||
NM_001190267.1 | NP_001177196.1 | 11 | 1337 | |||
NM_017974.3 | NP_060444.3 | 10 | 1284 | |||
NM_030803.6 | NP_110430.5 | 11 | 1341 | |||
NM_198890.2 | NP_942593.2 | 7 | 852 | |||
XM_005246082.2 | XP_005246139.1 | 12 | 1329 | |||
XM_005246084.2 | XP_005246141.1 | 9 | 868 | |||
XM_005246086.2 | XP_005246143.1 | 8 | 817 | |||
XM_006712608.2 | XP_006712671.1 | 11 | 1140 | |||
GenBank mRNA | AK000897.1 | - | 10 | 952 | ||
AK001690.1 | - | 10 | 1129 | |||
AK024453.1 | - | 10 | 1879 | |||
AK027854.1 | - | 9 | 866 | |||
AK308400.1 | - | 10 | 1182 | |||
AY358182.1 | - | 11 | 1230 | |||
AY398617.1 | - | 11 | 1230 | |||
BC071846.1 | - | 7 | 690 | |||
BC117337.1 | - | 11 | 1003 | |||
BC143900.1 | - | 10 | 946 | |||
EF079889.1 | - | 10 | 1027 | |||
EF079890.1 | - | 7 | 595 | |||
EF560734.1 | - | 10 | 1154 | |||
JQ924062.1 | - | 10 | 1027 |