Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018043.5 | NP_060513.5 | 9 | 1215 | ||
NR_030691.1 | 9 | 1063 | ||||
XM_006718602.2 | XP_006718665.1 | 10 | 997 | |||
XM_006718604.2 | XP_006718667.1 | 10 | 1112 | |||
XM_006718605.2 | XP_006718668.1 | 9 | 1058 | |||
XM_011545121.2 | XP_011543423.1 | 10 | 989 | |||
XM_011545123.2 | XP_011543425.1 | 10 | 989 | |||
XM_011545124.2 | XP_011543426.1 | 10 | 989 | |||
XM_011545125.2 | XP_011543427.1 | 10 | 989 | |||
XM_011545126.2 | XP_011543428.1 | 10 | 989 | |||
XM_011545127.2 | XP_011543429.1 | 9 | 923 | |||
XM_011545128.2 | XP_011543430.1 | 9 | 923 | |||
XM_011545129.2 | XP_011543431.1 | 10 | 904 | |||
XM_011545131.2 | XP_011543433.1 | 9 | 931 | |||
XM_017017956.1 | XP_016873445.1 | 9 | 923 | |||
XM_017017957.1 | XP_016873446.1 | 9 | 923 | |||
GenBank mRNA | AB845669.1 | - | 9 | 910 | ||
AK097619.1 | - | 9 | 931 | |||
AK293088.1 | - | 9 | 923 | |||
AL833582.1 | - | 9 | 959 | |||
AY728143.1 | - | 9 | 1215 | |||
BC027590.2 | - | 4 | 167 | |||
BC033036.2 | - | 9 | 1019 | 4823645 |