Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
siRNA ID
HSS182880
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
7
690
NM_001077352.1
NP_001070820.1
5
588
NM_001308044.1
NP_001294973.1
5
529
NM_018107.4
NP_060577.3
6
642
XM_011536890.2
XP_011535192.1
8
773
XM_011536892.2
XP_011535194.1
7
724
XM_011536893.2
XP_011535195.1
7
725
XM_011536894.2
XP_011535196.1
7
719
XM_011536895.2
XP_011535197.1
6
695
XM_011536896.2
XP_011535198.1
6
671
XM_011536897.2
XP_011535199.1
6
1057
XM_011536900.1
XP_011535202.1
6
633
XM_011536902.1
XP_011535204.1
6
539
XM_011536903.1
XP_011535205.1
4
415
XM_011536904.1
XP_011535206.1
5
472
XM_011536905.1
XP_011535207.1
4
470
XM_011536906.1
XP_011535208.1
3
361
XM_017021398.1
XP_016876887.1
7
727
XM_017021399.1
XP_016876888.1
6
674
XM_017021400.1
XP_016876889.1
6
592
XM_017021401.1
XP_016876890.1
7
687
XM_017021402.1
XP_016876891.1
7
598
XM_017021403.1
XP_016876892.1
6
616
XM_017021404.1
XP_016876893.1
6
566
XM_017021405.1
XP_016876894.1
6
633
XM_017021406.1
XP_016876895.1
7
1105
XM_017021407.1
XP_016876896.1
7
614
XM_017021408.1
XP_016876897.1
5
550
XM_017021409.1
XP_016876898.1
6
1051
XM_017021410.1
XP_016876899.1
5
572
XM_017021411.1
XP_016876900.1
5
1003
XM_017021412.1
XP_016876901.1
7
725
XM_017021413.1
XP_016876902.1
6
501
XM_017021414.1
XP_016876903.1
6
539
XM_017021415.1
XP_016876904.1
5
520
XM_017021416.1
XP_016876905.1
4
415
XM_017021417.1
XP_016876906.1
5
472
XM_017021418.1
XP_016876907.1
4
470
XM_017021419.1
XP_016876908.1
3
361
XM_017021420.1
XP_016876909.1
4
890
GenBank mRNA
AF087905.1
-
6
593
AF275678.1
-
7
645
AK001344.1
-
5
538
AK125593.1
-
7
652
AK225792.1
-
7
652
AK303668.1
-
5
491
AK308055.1
-
6
605
AK315620.1
-
6
604
AL834198.1
-
7
651
BC002566.2
-
6
620
3138909
BC024208.2
-
6
579
2960467
BC106012.1
-
4
389
BX161440.1
-
7
623
CK000110.1
-
6
616
CK003276.1
-
5
521
CR457257.1
-
5
443
KU178687.1
-
5
443
KU178688.1
-
6
491
KU178689.1
-
4
389
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0