Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF10L SIRNA Assays ›
Gene Symbol:
ARHGEF10L
Gene Name:
Rho guanine nucleotide exchange factor 10 like
Gene Aliases:
GrinchGEF
Chromosome Location:
Chr.1: 17520197 - 17697875 on Build GRCh38
UniGene ID:
Hs.443460
Species:
Human
siRNA ID
HSS182891
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF10L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011722.2
NP_001011722.2
27
3543
NM_001319837.1
NP_001306766.1
26
3528
NM_001319838.1
NP_001306767.1
18
2767
NM_001328124.1
NP_001315053.1
21
2992
NM_018125.3
NP_060595.3
29
3776
NR_137287.1
30
4523
NR_137288.1
29
4403
XM_005245923.2
XP_005245980.2
28
3635
XM_005245925.2
XP_005245982.2
28
3623
XM_005245927.2
XP_005245984.2
21
2992
XM_005245929.2
XP_005245986.2
20
2977
XM_006710728.1
XP_006710791.1
28
3817
XM_006710729.1
XP_006710792.1
28
3632
XM_006710731.1
XP_006710794.1
28
3814
XM_011541691.2
XP_011539993.1
27
3632
XM_011541692.2
XP_011539994.1
26
3626
XM_011541693.2
XP_011539995.1
26
3617
XM_017001617.1
XP_016857106.1
27
3608
XM_017001618.1
XP_016857107.1
22
2910
XM_017001621.1
XP_016857110.1
28
3759
XM_017001622.1
XP_016857111.1
27
3744
GenBank mRNA
AB046846.1
-
29
3776
AK001383.1
-
17
2307
AK092020.1
-
18
2764
AK123471.1
-
2
1544
AK126877.1
-
20
3882
AK300264.1
-
20
2940
BC065561.1
-
15
2278
BC080596.1
-
18
2439
BC117171.1
-
27
3533
40125722
BC143347.1
-
27
3533
9051852
DQ139833.1
-
26
3485
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
positive regulation of GTPase activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline