Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297760.1 | NP_001284689.1 | 4 | 1079 | ||
NM_001297761.1 | NP_001284690.1 | 3 | 819 | |||
NM_001297762.1 | NP_001284691.1 | 3 | 956 | |||
NM_018208.3 | NP_060678.2 | 4 | 1079 | |||
XM_011509714.2 | XP_011508016.1 | 3 | 319 | |||
XM_011509715.1 | XP_011508017.1 | 3 | 624 | |||
XM_017001632.1 | XP_016857121.1 | 3 | 407 | |||
GenBank mRNA | AB073608.1 | - | 4 | 679 | ||
AK001623.1 | - | 4 | 791 | |||
AK293426.1 | - | 3 | 527 | |||
AK302145.1 | - | 3 | 556 | |||
AK316493.1 | - | 3 | 315 | |||
BC010082.2 | - | 4 | 900 | 4329734 |