Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAVER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018211.3 | NP_060681.2 | 4 | 953 | ||
XM_006710738.3 | XP_006710801.2 | 4 | 1060 | |||
XM_011541706.2 | XP_011540008.1 | 4 | 1069 | |||
GenBank mRNA | AB046799.1 | - | 4 | 953 | ||
AK001632.1 | - | 2 | 116 | |||
AK299228.1 | - | 2 | 124 | |||
BC065303.1 | - | 2 | 344 |