Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018283.3 | NP_060753.1 | 3 | 615 | ||
NR_136687.1 | 3 | 615 | ||||
NR_136688.1 | 3 | 615 | ||||
GenBank mRNA | AK001818.1 | - | 3 | 615 | ||
BC064607.1 | - | 3 | 434 | |||
BC107875.1 | - | 3 | 488 | 6451308 | ||
BC133015.1 | - | 3 | 556 | 40147037 | ||
BC133017.1 | - | 3 | 556 | 40147039 | ||
DA000395.1 | - | 3 | 400 | |||
DA760263.1 | - | 3 | 421 |