Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104629.1 | NP_001098099.1 | 4 | 909 | ||
NM_018302.2 | NP_060772.2 | 3 | 904 | |||
XM_011513712.2 | XP_011512014.1 | 5 | 973 | |||
XM_011513713.2 | XP_011512015.1 | 4 | 1175 | |||
XM_017008361.1 | XP_016863850.1 | 2 | 2105 | |||
GenBank mRNA | AK001879.1 | - | 3 | 904 | ||
AK225192.1 | - | 3 | 873 | |||
BC037906.1 | - | 4 | 870 | 5276648 |