Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018317.3 | NP_060787.2 | 2 | 382 | ||
XM_011513852.2 | XP_011512154.1 | 2 | 222 | |||
XM_017008363.1 | XP_016863852.1 | 2 | 222 | |||
XM_017008364.1 | XP_016863853.1 | 2 | 222 | |||
GenBank mRNA | AK001944.1 | - | 2 | 222 | ||
AK225395.1 | - | 2 | 222 | |||
AK289631.1 | - | 2 | 222 | |||
AK310795.1 | - | 2 | 229 | |||
BC031642.1 | - | 2 | 178 | 5175317 | ||
DC299740.1 | - | 2 | 382 |