Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018320.4 | NP_060790.2 | 7 | 983 | ||
NR_024147.1 | 6 | 945 | ||||
NR_024148.1 | 7 | 1083 | ||||
XM_006718629.1 | XP_006718692.1 | 6 | 896 | |||
XM_006718630.1 | XP_006718693.1 | 8 | 796 | |||
XM_006718632.1 | XP_006718695.1 | 5 | 741 | |||
XM_011545150.1 | XP_011543452.1 | 8 | 828 | |||
GenBank mRNA | AK001961.1 | - | 2 | 147 | ||
AK023139.1 | - | 7 | 649 | |||
AK094508.1 | - | 7 | 749 | |||
BC009672.2 | - | 7 | 651 | |||
BC063680.1 | - | 2 | 159 | |||
BC093084.1 | - | 6 | 585 | |||
BC117273.1 | - | 7 | 615 | |||
BX463021.2 | - | 3 | 250 | |||
CR457298.1 | - | 7 | 643 |