Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099455.1 | NP_001092925.1 | 3 | 532 | ||
NM_018340.2 | NP_060810.2 | 4 | 958 | |||
GenBank mRNA | AK002013.1 | - | 4 | 941 | ||
AK022710.1 | - | 4 | 908 | |||
AK093854.1 | - | 4 | 704 | |||
AK225359.1 | - | 4 | 908 | |||
AK225617.1 | - | 4 | 953 | |||
AK298662.1 | - | 3 | 362 | |||
AY553877.1 | - | 4 | 847 | |||
BC006289.2 | - | 4 | 942 | 3959701 | ||
BX640805.1 | - | 3 | 498 |