Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018346.2 | NP_060816.1 | 4 | 574 | ||
NR_130911.1 | 2 | 235 | ||||
GenBank mRNA | AK002026.1 | - | 4 | 545 | ||
AK022105.1 | - | 4 | 514 | |||
AK222472.1 | - | 4 | 525 | |||
AK297655.1 | - | 2 | 175 | |||
BC005854.2 | - | 3 | 326 | |||
BC050538.1 | - | 4 | 505 | 5214885 |