Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 3 | 852 | ||
NR_130759.1 | 2 | 641 | ||||
NR_130760.1 | 3 | 852 | ||||
XM_011518839.1 | XP_011517141.1 | 3 | 852 | |||
XM_017014889.1 | XP_016870378.1 | 3 | 804 | |||
GenBank mRNA | AK002137.1 | - | 3 | 457 | ||
AK310397.1 | - | 2 | 197 | |||
BC005202.1 | - | 3 | 385 | 3933738 | ||
BC017914.1 | - | 3 | 423 | 4292375 | ||
BX647652.1 | - | 3 | 424 |