Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 3 | 187 | ||
GenBank mRNA | AB036706.1 | - | 3 | 99 | ||
AF271386.1 | - | 3 | 110 | |||
AK000029.1 | - | 3 | 136 | |||
AK222533.1 | - | 3 | 174 | |||
AY065972.1 | - | 3 | 177 | |||
AY157361.1 | - | 3 | 189 | |||
AY157362.1 | - | 3 | 181 | |||
AY358359.1 | - | 3 | 173 | |||
AY549722.1 | - | 3 | 276 | |||
AY619691.1 | - | 3 | 97 | |||
AY619692.1 | - | 3 | 154 | |||
BC020664.1 | - | 3 | 187 | 4693329 |