Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034833.1 | NP_001030005.1 | 2 | 341 | ||
NM_017838.3 | NP_060308.1 | 2 | 341 | |||
GenBank mRNA | AF161404.1 | - | 2 | 251 | ||
AF401219.1 | - | 2 | 199 | |||
AJ293309.1 | - | 2 | 265 | |||
AK000486.1 | - | 2 | 247 | |||
BC000009.2 | - | 2 | 249 | 3505294 | ||
BC006387.2 | - | 2 | 275 | 4109843 | ||
BE797477.1 | - | 2 | 361 | |||
BG827491.1 | - | 2 | 349 | |||
BM913344.1 | - | 2 | 241 | |||
CR457238.1 | - | 2 | 198 |