Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSDMB SIRNA Assays ›
Gene Symbol:
GSDMB
Gene Name:
gasdermin B
Gene Aliases:
GSDML, PP4052, PRO2521
Chromosome Location:
Chr.17: 39904595 - 39919377 on Build GRCh38
UniGene ID:
Hs.306777
Species:
Human
siRNA ID
HSS183332
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSDMB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042471.1
NP_001035936.1
7
956
NM_001165958.1
NP_001159430.1
8
995
NM_001165959.1
NP_001159431.1
7
959
NM_018530.2
NP_061000.2
5
1022
XM_011525004.1
XP_011523306.1
8
2403
XM_011525006.1
XP_011523308.1
8
2394
XM_011525007.2
XP_011523309.1
8
1725
XM_011525009.1
XP_011523311.1
8
2083
XM_011525011.1
XP_011523313.1
8
1734
XM_011525012.1
XP_011523314.1
8
2074
XM_011525013.1
XP_011523315.1
9
1792
XM_011525015.1
XP_011523317.1
9
1783
XM_017024848.1
XP_016880337.1
8
2726
XM_017024849.1
XP_016880338.1
9
2435
XM_017024850.1
XP_016880339.1
7
2925
XM_017024851.1
XP_016880340.1
6
2910
XM_017024852.1
XP_016880341.1
6
2886
GenBank mRNA
AF119884.1
-
5
1022
AF258572.1
-
1
1173
AK000409.1
-
7
896
AK057588.1
-
1
1605
AK296607.1
-
7
944
BC025682.1
-
6
872
5205060
BX538068.1
-
7
659
BX647700.1
-
6
4932
CA488357.1
-
3
376
EU526753.1
-
6
825
EU526754.1
-
4
713
EU526755.1
-
5
740
EU526756.1
-
4
571
Back To Top
More Information
Gene Ontology Categories:
Function(s)
programmed cell death
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0