Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292039.1 | NP_001278968.1 | 5 | 1200 | ||
NM_001292040.1 | NP_001278969.1 | 4 | 2240 | |||
NM_002747.3 | NP_002738.2 | 6 | 2616 | |||
XM_005258299.3 | XP_005258356.1 | 6 | 6978 | |||
XM_011526074.2 | XP_011524376.1 | 6 | 7160 | |||
XM_011526075.2 | XP_011524377.1 | 6 | 2618 | |||
XM_011526076.2 | XP_011524378.1 | 6 | 2981 | |||
XM_011526077.2 | XP_011524379.1 | 5 | 3627 | |||
XM_011526078.2 | XP_011524380.1 | 4 | 1732 | |||
XM_017025839.1 | XP_016881328.1 | 6 | 2549 | |||
GenBank mRNA | AK293818.1 | - | 5 | 1181 | ||
AK295058.1 | - | 6 | 2537 | |||
AK303601.1 | - | 5 | 1278 | |||
BC040493.1 | - | 4 | 2212 | |||
BC050299.1 | - | 6 | 2616 | 5303135 | ||
BC117216.1 | - | 5 | 1670 | 40125767 | ||
BC143769.1 | - | 3 | 1292 | |||
X59727.1 | - | 5 | 2148 |