Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
siRNA ID
HSS183385
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
6
577
NM_001318068.1
NP_001304997.1
6
672
NM_001318069.1
NP_001304998.1
7
1142
NM_002753.4
NP_002744.1
7
678
NM_138980.3
NP_620446.1
7
1169
NM_138982.3
NP_620448.1
7
1142
XM_005263129.2
XP_005263186.1
7
1239
XM_005263130.2
XP_005263187.1
7
635
XM_005263131.2
XP_005263188.1
7
1036
XM_005263135.3
XP_005263192.1
7
1036
XM_006714268.2
XP_006714331.1
7
909
XM_006714269.2
XP_006714332.1
7
1036
XM_011532117.2
XP_011530419.1
7
742
XM_011532118.2
XP_011530420.1
7
835
XM_011532120.2
XP_011530422.1
7
805
XM_011532121.2
XP_011530423.1
7
671
XM_017008419.1
XP_016863908.1
8
932
XM_017008420.1
XP_016863909.1
7
1366
XM_017008421.1
XP_016863910.1
7
962
XM_017008422.1
XP_016863911.1
5
462
XM_017008423.1
XP_016863912.1
7
654
XM_017008424.1
XP_016863913.1
7
989
XM_017008425.1
XP_016863914.1
7
662
XM_017008426.1
XP_016863915.1
7
990
XM_017008427.1
XP_016863916.1
7
1063
XM_017008428.1
XP_016863917.1
7
805
XM_017008429.1
XP_016863918.1
7
1366
XM_017008430.1
XP_016863919.1
7
744
XM_017008431.1
XP_016863920.1
7
962
XM_017008432.1
XP_016863921.1
7
635
XM_017008433.1
XP_016863922.1
7
1063
XM_017008434.1
XP_016863923.1
7
1019
XM_017008435.1
XP_016863924.1
7
909
XM_017008436.1
XP_016863925.1
7
1393
XM_017008437.1
XP_016863926.1
7
671
XM_017008438.1
XP_016863927.1
7
662
XM_017008439.1
XP_016863928.1
7
990
XM_017008440.1
XP_016863929.1
7
1063
XM_017008441.1
XP_016863930.1
7
1063
XM_017008442.1
XP_016863931.1
9
1384
XM_017008443.1
XP_016863932.1
9
1035
XM_017008444.1
XP_016863933.1
9
1357
XM_017008445.1
XP_016863934.1
6
866
XM_017008446.1
XP_016863935.1
4
1558
XM_017008447.1
XP_016863936.1
6
1004
XM_017008448.1
XP_016863937.1
6
977
XM_017008449.1
XP_016863938.1
6
1004
XM_017008450.1
XP_016863939.1
6
977
XM_017008451.1
XP_016863940.1
6
803
XM_017008452.1
XP_016863941.1
6
1004
XM_017008453.1
XP_016863942.1
5
462
XM_017008454.1
XP_016863943.1
7
742
GenBank mRNA
AK057723.1
-
2
464
AK124791.1
-
7
1394
AK225974.1
-
7
723
AK294125.1
-
3
193
AK307129.1
-
7
796
BC022492.1
-
7
975
BC035057.1
-
7
987
5261221
BC051731.1
-
7
788
4838495
BC065516.1
-
2
139
5721954
U07620.1
-
7
679
U34819.1
-
6
523
U34820.1
-
6
547
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline