Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135651.2 | NP_001129123.1 | 10 | 1064 | ||
NM_001135652.2 | NP_001129124.1 | 8 | 757 | |||
NM_002759.3 | NP_002750.1 | 10 | 1298 | |||
XM_011532987.2 | XP_011531289.1 | 10 | 998 | |||
XM_017004503.1 | XP_016859992.1 | 10 | 994 | |||
GenBank mRNA | AK290655.1 | - | 10 | 984 | ||
AK313818.1 | - | 9 | 821 | |||
AY302136.1 | - | 8 | 741 | |||
BC007769.2 | - | 9 | 840 | |||
BC022314.1 | - | 10 | 952 | |||
BC040851.1 | - | 10 | 1064 | |||
BC057805.1 | - | 10 | 969 | |||
BC093676.1 | - | 8 | 769 | 7939521 | ||
BC101475.1 | - | 8 | 769 | 8068981 | ||
BC143228.1 | - | 8 | 768 | |||
BC143230.1 | - | 8 | 769 | 9051731 | ||
M35663.1 | - | 9 | 927 | |||
M85294.1 | - | 8 | 771 |