Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000948.5 | NP_000939.1 | 4 | 876 | ||
NM_001163558.2 | NP_001157030.1 | 5 | 548 | |||
XM_011514753.2 | XP_011513055.1 | 5 | 595 | |||
XM_011514754.2 | XP_011513056.1 | 6 | 608 | |||
GenBank mRNA | BC015850.1 | - | 4 | 879 | 4657724 | |
BC088370.1 | - | 4 | 406 | 30393428 | ||
D00411.1 | - | 3 | 329 | |||
DQ839492.1 | - | 5 | 264 | |||
GQ305133.1 | - | 5 | 548 | |||
M29386.1 | - | 3 | 329 | |||
V00566.1 | - | 4 | 361 | |||
X54393.1 | - | 5 | 498 |