Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306188.1 | NP_001293117.1 | 2, 3 | 407 | ||
NM_019556.2 | NP_062456.1 | 2, 3 | 407 | |||
XM_011531366.2 | XP_011529668.1 | 2, 3 | 361 | |||
XM_017029653.1 | XP_016885142.1 | 3, 4 | 600 | |||
XM_017029654.1 | XP_016885143.1 | 2, 3 | 361 | |||
GenBank mRNA | AK299074.1 | - | 2, 3 | 320 | ||
AK299172.1 | - | 1, 2 | 197 | |||
AK316570.1 | - | 2, 3 | 282 | |||
AL137163.1 | - | 2, 3 | 330 | |||
BC005700.1 | - | 2, 3 | 310 | 3450155 | ||
CB997365.1 | - | 2, 3 | 315 | |||
CN428237.1 | - | 2, 3 | 409 | |||
DQ323992.1 | - | 1, 2 | 143 |