Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021800.2 | NP_068572.1 | 1, 2 | 235 | ||
NM_201262.1 | NP_957714.1 | 1, 2 | 235 | |||
GenBank mRNA | AF176012.1 | - | 1, 2 | 220 | ||
AF176013.1 | - | 1, 2 | 152 | |||
AK289537.1 | - | 1, 2 | 185 | |||
BC017018.1 | - | 1, 2 | 171 | 3884093 | ||
CB121724.1 | - | 1, 2 | 237 | |||
CR457184.1 | - | 1, 2 | 67 |